Assay Details
Gene Symbol: wee2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103029.1 | NP_001096499.1 | 648 | 73 | ||
XM_012959340.2 | XP_012814794.1 | 725 | 73 | |||
GenBank mRNA | BC135215.1 | - | 648 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
