Assay Details
Gene Symbol: mrpl46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103031.1 | NP_001096501.1 | 1 - 2 | 237 | 68 | |
GenBank mRNA | BC135281.1 | - | 1 - 2 | 237 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
