Assay Details
Gene Symbol: st6galnac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204083.2 | NP_989414.2 | 6 - 7 | 903 | 65 | |
XM_018097688.1 | XP_017953177.1 | 7 - 8 | 1349 | 65 | ||
GenBank mRNA | AJ620650.1 | - | 6 - 7 | 872 | 65 | |
BC135965.1 | - | 6 - 7 | 903 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
