Assay Details
Gene Symbol: arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016131.5 | NP_001016131.1 | 3 - 4 | 392 | 78 | |
GenBank mRNA | BC118695.2 | - | 3 - 4 | 393 | 78 | |
CR762046.2 | - | 3 - 4 | 304 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
