Assay Details
Gene Symbol: tmem218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016711.3 | NP_001016711.1 | 2 - 3 | 223 | 74 | |
XM_012966618.2 | XP_012822072.1 | 2 - 3 | 279 | 74 | ||
XM_012966620.2 | XP_012822074.1 | 2 - 3 | 457 | 74 | ||
GenBank mRNA | BC158955.1 | - | 2 - 3 | 223 | 74 | |
CR760114.2 | - | 1 - 2 | 170 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
