Assay Details
| Gene Symbol: bcl7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103072.1 | NP_001096542.1 | 5 - 6 | 504 | 64 | |
| XM_012956320.2 | XP_012811774.1 | 5 - 6 | 615 | 64 | ||
| GenBank mRNA | BC136023.1 | - | 5 - 6 | 504 | 64 | |
More Information
Set Membership: |
|

bcl7b
Genomic Map