Assay Details
| Gene Symbol: nacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103074.1 | NP_001096544.1 | 4 - 5 | 1714 | 85 | |
| XM_012967897.2 | XP_012823351.1 | 4 - 5 | 1263 | 85 | ||
| XM_012967898.2 | XP_012823352.1 | 4 - 5 | 1308 | 85 | ||
| GenBank mRNA | BC136033.1 | - | 4 - 5 | 1714 | 85 | |
More Information
Set Membership: |
|

nacc2
Genomic Map