Assay Details
Gene Symbol: chst11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103076.1 | NP_001096546.1 | 2 - 3 | 732 | 60 | |
XM_012958471.2 | XP_012813925.1 | 2 - 3 | 959 | 60 | ||
GenBank mRNA | BC136103.1 | - | 2 - 3 | 732 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
