Assay Details
| Gene Symbol: acod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103172.1 | NP_001096642.1 | 2 - 3 | 132 | 98 | |
| GenBank mRNA | CT030574.1 | - | 2 - 3 | 132 | 98 | |
| Gene Symbol: LOC100490060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002931800.4 | XP_002931846.1 | 2 - 3 | 165 | 98 | |
More Information
Set Membership: |
|

acod1
Genomic Map