Assay Details
| Gene Symbol: acod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103172.1 | NP_001096642.1 | 5 - 6 | 629 | 67 | |
| XM_018096124.1 | XP_017951613.1 | 4 - 5 | 587 | 67 | ||
| GenBank mRNA | CT030574.1 | - | 5 - 6 | 629 | 67 | |
| Gene Symbol: LOC100490060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002931800.4 | XP_002931846.1 | 5 - 6 | 662 | 67 | |
| XM_018096602.1 | XP_017952091.1 | 4 - 5 | 589 | 67 | ||
More Information
Set Membership: |
|

acod1
Genomic Map