Assay Details
Gene Symbol: sec61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110044.1 | NP_001103514.1 | 3 - 4 | 294 | 66 | |
GenBank mRNA | BC135617.2 | - | 3 - 4 | 294 | 66 | |
CR942631.1 | - | 3 - 4 | 230 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
