Assay Details
Gene Symbol: prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012961768.2 | XP_012817222.1 | 2 - 3 | 796 | 72 | |
XM_012961769.1 | XP_012817223.1 | 2 - 3 | 438 | 72 | ||
XM_012961771.2 | XP_012817225.1 | 2 - 3 | 273 | 72 | ||
GenBank mRNA | BC153335.1 | - | 2 - 3 | 545 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
