Assay Details
Gene Symbol: tcf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110048.1 | NP_001103518.1 | 1 - 2 | 556 | 107 | |
GenBank mRNA | BC153711.1 | - | 1 - 2 | 556 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
