Assay Details
Gene Symbol: leap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112914.1 | NP_001106385.1 | 2 - 3 | 197 | 81 | |
GenBank mRNA | BC155465.1 | - | 2 - 3 | 197 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
