Assay Details
| Gene Symbol: acer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112961.2 | NP_001106432.1 | 4 - 5 | 469 | 66 | |
| XM_012969733.2 | XP_012825187.1 | 4 - 5 | 688 | 66 | ||
| GenBank mRNA | EL802545.1 | - | 4 - 5 | 493 | 66 | |
| EL802546.1 | - | 3 - 4 | 605 | 66 | ||
More Information
Set Membership: |
|

acer1
Genomic Map