Assay Details
Gene Symbol: lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112972.1 | NP_001106443.1 | 2 - 3 | 153 | 93 | |
GenBank mRNA | BC153732.1 | - | 2 - 3 | 153 | 93 |
Gene Symbol: lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018094389.1 | XP_017949878.1 | 1 - 2 | 93 | ||
XM_018094390.1 | XP_017949879.1 | 1 - 2 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
