Assay Details
Gene Symbol: naa60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112973.1 | NP_001106444.1 | 8 - 9 | 1052 | 67 | |
XM_012969997.2 | XP_012825451.1 | 7 - 8 | 1056 | 67 | ||
GenBank mRNA | BC153734.1 | - | 8 - 9 | 1052 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
