Assay Details
Gene Symbol: oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112991.1 | NP_001106462.1 | 15 - 16 | 2660 | 72 | |
XM_012964221.1 | XP_012819675.1 | 15 - 16 | 2774 | 72 | ||
XM_012964222.1 | XP_012819676.1 | 15 - 16 | 2774 | 72 | ||
XM_012964223.1 | XP_012819677.1 | 13 - 14 | 2504 | 72 | ||
XM_012964224.1 | XP_012819678.1 | 14 - 15 | 2693 | 72 | ||
XM_012964225.1 | XP_012819679.1 | 14 - 15 | 2693 | 72 | ||
XM_012964226.2 | XP_012819680.1 | 12 - 13 | 2422 | 72 | ||
XM_012964227.2 | XP_012819681.1 | 5 - 6 | 869 | 72 | ||
XM_012964228.2 | XP_012819682.1 | 5 - 6 | 867 | 72 | ||
XM_012964229.2 | XP_012819683.1 | 4 - 5 | 764 | 72 | ||
XM_012964230.2 | XP_012819684.1 | 4 - 5 | 710 | 72 | ||
XM_018094522.1 | XP_017950011.1 | 15 - 16 | 2774 | 72 | ||
GenBank mRNA | BC154076.1 | - | 15 - 16 | 2660 | 72 | |
BC159172.1 | - | 12 - 13 | 2343 | 72 | ||
CU075896.1 | - | 4 - 5 | 511 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
