Assay Details
| Gene Symbol: tmigd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113059.1 | NP_001106530.1 | 6 - 7 | 863 | 86 | |
| GenBank mRNA | BC154899.1 | - | 6 - 7 | 863 | 86 | |
More Information
Set Membership: |
|

tmigd1
Genomic Map