Assay Details
| Gene Symbol: xrcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113080.2 | NP_001106551.1 | 2 - 3 | 231 | 87 | |
| XM_012970771.2 | XP_012826225.1 | 2 - 3 | 284 | 87 | ||
| XM_012970791.2 | XP_012826245.1 | 2 - 3 | 284 | 87 | ||
| GenBank mRNA | BC154929.1 | - | 2 - 3 | 231 | 87 | |
More Information
Set Membership: |
|

xrcc4
Genomic Map