Assay Details
Gene Symbol: ltc4s.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113094.1 | NP_001106565.1 | 2 - 3 | 333 | 75 | |
GenBank mRNA | BC155013.1 | - | 2 - 3 | 333 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
