Assay Details
| Gene Symbol: inka2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113117.1 | NP_001106588.1 | 1 - 2 | 173 | 62 | |
| GenBank mRNA | BC155411.1 | - | 1 - 2 | 173 | 62 | |
| CR761080.2 | - | 1 - 2 | 385 | 62 | ||
More Information
Set Membership: |
|

inka2
Genomic Map