Assay Details
Gene Symbol: npy4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113121.1 | NP_001106592.1 | 1 | 1148 | 100 | |
XM_012958511.2 | XP_012813965.1 | 2 | 2583 | 100 | ||
GenBank mRNA | BC155422.1 | - | 1 | 1148 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
