Assay Details
Gene Symbol: aplf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113131.1 | NP_001106602.1 | 2 - 3 | 242 | 75 | |
XM_012962661.2 | XP_012818115.1 | 2 - 3 | 264 | 75 | ||
XM_012962662.2 | XP_012818116.1 | 2 - 3 | 315 | 75 | ||
GenBank mRNA | BC155435.1 | - | 2 - 3 | 242 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
