Assay Details
Gene Symbol: mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113157.1 | NP_001106628.1 | 2 - 3 | 324 | 88 | |
GenBank mRNA | BC155510.1 | - | 2 - 3 | 324 | 88 | |
CU075317.1 | - | 2 - 3 | 324 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
