Assay Details
| Gene Symbol: slc1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113216.1 | NP_001106687.1 | 3 - 4 | 475 | 70 | |
| GenBank mRNA | BC084737.1 | - | 3 - 4 | 479 | 70 | |
| BC154714.1 | - | 3 - 4 | 475 | 70 | ||
More Information
Set Membership: |
|

slc1a3
Genomic Map