Assay Details
Gene Symbol: bloc1s5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113668.1 | NP_001107140.1 | 3 - 4 | 416 | 63 | |
GenBank mRNA | BC157360.1 | - | 3 - 4 | 416 | 63 | |
BC170587.1 | - | 3 - 4 | 416 | 63 | ||
BC170589.1 | - | 3 - 4 | 416 | 63 | ||
CT030090.1 | - | 3 - 4 | 362 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
