Assay Details
Gene Symbol: map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113685.1 | NP_001107157.1 | 8 - 9 | 833 | 52 | |
GenBank mRNA | BC157556.1 | - | 8 - 9 | 833 | 52 | |
BC170945.1 | - | 8 - 9 | 833 | 52 | ||
BC170947.1 | - | 8 - 9 | 833 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
