Assay Details
Gene Symbol: mall | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113808.1 | NP_001107280.1 | 2 - 3 | 328 | 98 | |
GenBank mRNA | BC087766.1 | - | 2 - 3 | 314 | 98 | |
BC122087.1 | - | 2 - 3 | 308 | 98 | ||
BC154915.1 | - | 2 - 3 | 328 | 98 | ||
BC169149.1 | - | 2 - 3 | 297 | 98 | ||
BC171228.1 | - | 2 - 3 | 314 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
