Assay Details
Gene Symbol: bicra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113827.1 | NP_001107299.1 | 15 - 16 | 4205 | 66 | |
XM_012967954.2 | XP_012823408.1 | 13 - 14 | 4633 | 66 | ||
XM_012967955.2 | XP_012823409.1 | 13 - 14 | 4421 | 66 | ||
XM_018096136.1 | XP_017951625.1 | 14 - 15 | 4704 | 66 | ||
GenBank mRNA | BC155694.1 | - | 15 - 16 | 4205 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
