Assay Details
Gene Symbol: bicra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113827.1 | NP_001107299.1 | 8 - 9 | 2676 | 67 | |
XM_012967954.2 | XP_012823408.1 | 6 - 7 | 3077 | 67 | ||
XM_018096136.1 | XP_017951625.1 | 7 - 8 | 3175 | 67 | ||
GenBank mRNA | BC155694.1 | - | 8 - 9 | 2676 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
