Assay Details
Gene Symbol: tex261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113851.1 | NP_001107323.1 | 1 - 2 | 386 | 61 | |
GenBank mRNA | BC155995.1 | - | 1 - 2 | 386 | 61 | |
BC170954.1 | - | 1 - 2 | 191 | 61 | ||
BC170958.1 | - | 1 - 2 | 191 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
