Assay Details
Gene Symbol: rpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113853.1 | NP_001107325.1 | 1 - 2 | 154 | 71 | |
XM_012953424.1 | XP_012808878.1 | 3 - 4 | 662 | 71 | ||
GenBank mRNA | BC156002.1 | - | 1 - 2 | 154 | 71 | |
CU075433.1 | - | 1 - 2 | 150 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
