Assay Details
Gene Symbol: fmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113864.1 | NP_001107336.1 | 1 - 2 | 253 | 92 | |
GenBank mRNA | BC157136.1 | - | 1 - 2 | 253 | 92 | |
BC171278.1 | - | 1 - 2 | 238 | 92 | ||
BC171284.1 | - | 1 - 2 | 216 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
