Assay Details
Gene Symbol: fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113881.1 | NP_001107353.1 | 2 - 3 | 259 | 79 | |
XM_012960561.2 | XP_012816015.1 | 2 - 3 | 323 | 79 | ||
XM_012960562.2 | XP_012816016.1 | 2 - 3 | 455 | 79 | ||
GenBank mRNA | BC157195.1 | - | 2 - 3 | 259 | 79 | |
BC171197.1 | - | 2 - 3 | 236 | 79 | ||
BC171199.1 | - | 2 - 3 | 236 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
