Assay Details
Gene Symbol: ucp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113882.1 | NP_001107354.1 | 5 - 6 | 673 | 100 | |
GenBank mRNA | BC157201.1 | - | 5 - 6 | 673 | 100 | |
BC171056.1 | - | 4 - 5 | 660 | 100 | ||
BC171062.1 | - | 4 - 5 | 660 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
