Assay Details
| Gene Symbol: hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113890.1 | NP_001107362.1 | 2 - 3 | 441 | 58 | |
| XM_012960563.2 | XP_012816017.1 | 3 - 4 | 987 | 58 | ||
| XM_012960564.2 | XP_012816018.1 | 3 - 4 | 1035 | 58 | ||
| XM_012960565.1 | XP_012816019.1 | 3 - 4 | 508 | 58 | ||
| GenBank mRNA | BC082746.1 | - | 3 - 4 | 510 | 58 | |
| BC157213.1 | - | 2 - 3 | 441 | 58 | ||
| BC171074.1 | - | 2 - 3 | 334 | 58 | ||
| BC171076.1 | - | 2 - 3 | 334 | 58 | ||
More Information
Set Membership: |
|

hyi
Genomic Map