Assay Details
Gene Symbol: izumo1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113896.1 | NP_001107368.1 | 1 - 2 | 142 | 93 | |
XM_012960567.1 | XP_012816021.1 | 2 - 3 | 231 | 93 | ||
GenBank mRNA | BC157228.1 | - | 1 - 2 | 142 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
