Assay Details
Gene Symbol: izumo1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113896.1 | NP_001107368.1 | 5 - 6 | 714 | 74 | |
XM_012960567.1 | XP_012816021.1 | 6 - 7 | 803 | 74 | ||
GenBank mRNA | BC157228.1 | - | 5 - 6 | 714 | 74 | |
BC161296.1 | - | 5 - 6 | 735 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
