Assay Details
Gene Symbol: sra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113899.1 | NP_001107371.1 | 524 | 63 | ||
GenBank mRNA | BC157234.1 | - | 524 | 63 | ||
BC171161.1 | - | 304 | 63 | |||
BC171163.1 | - | 304 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
