Assay Details
| Gene Symbol: ramp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113904.1 | NP_001107376.1 | 1 - 2 | 363 | 97 | |
| GenBank mRNA | BC157246.1 | - | 1 - 2 | 363 | 97 | |
| BC170966.1 | - | 1 - 2 | 228 | 97 | ||
| BC170968.1 | - | 1 - 2 | 228 | 97 | ||
More Information
Set Membership: |
|

ramp3
Genomic Map