Assay Details
| Gene Symbol: rccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113913.1 | NP_001107385.1 | 5 - 6 | 995 | 96 | |
| XM_012958528.2 | XP_012813982.1 | 6 - 7 | 2342 | 96 | ||
| GenBank mRNA | BC157279.1 | - | 5 - 6 | 995 | 96 | |
More Information
Set Membership: |
|

rccd1
Genomic Map