Assay Details
Gene Symbol: wbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113925.1 | NP_001107397.1 | 2 - 3 | 209 | 80 | |
GenBank mRNA | BC157344.1 | - | 2 - 3 | 209 | 80 | |
BC158898.1 | - | 2 - 3 | 209 | 80 | ||
CT027883.2 | - | 2 - 3 | 208 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
