Assay Details
Gene Symbol: sgo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114031.1 | NP_001107503.1 | 8 - 9 | 1701 | 77 | |
XM_012964257.2 | XP_012819711.1 | 1761 | 77 | |||
GenBank mRNA | BC157661.1 | - | 8 - 9 | 1701 | 77 | |
CR926350.2 | - | 1660 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
