Assay Details
Gene Symbol: fsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114044.1 | NP_001107516.1 | 8 - 9 | 875 | 68 | |
GenBank mRNA | BC157680.1 | - | 8 - 9 | 875 | 68 | |
BC171207.1 | - | 8 - 9 | 862 | 68 | ||
BC171211.1 | - | 8 - 9 | 862 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
