Assay Details
Gene Symbol: mre11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114047.1 | NP_001107519.1 | 15 - 16 | 1887 | 61 | |
XM_012956401.2 | XP_012811855.1 | 16 - 17 | 1964 | 61 | ||
XM_012956402.2 | XP_012811856.1 | 16 - 17 | 1986 | 61 | ||
GenBank mRNA | BC157691.1 | - | 15 - 16 | 1887 | 61 | |
BC171212.1 | - | 15 - 16 | 1878 | 61 | ||
BC171218.1 | - | 15 - 16 | 1878 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
