Assay Details
Gene Symbol: pdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114067.1 | NP_001107539.1 | 2 - 3 | 123 | 97 | |
GenBank mRNA | BC157793.1 | - | 2 - 3 | 123 | 97 | |
BC171294.1 | - | 2 - 3 | 123 | 97 | ||
BC171298.1 | - | 2 - 3 | 123 | 97 | ||
CR942709.2 | - | 2 - 3 | 125 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
