Assay Details
| Gene Symbol: pmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114072.1 | NP_001107544.1 | 1 - 2 | 98 | 64 | |
| GenBank mRNA | BC157804.1 | - | 1 - 2 | 98 | 64 | |
| BC171082.1 | - | 1 - 2 | 98 | 64 | ||
| BC171084.1 | - | 1 - 2 | 98 | 64 | ||
| CU075393.1 | - | 1 - 2 | 80 | 64 | ||
More Information
Set Membership: |
|

pmp2
Genomic Map