Assay Details
| Gene Symbol: gnrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114077.1 | NP_001107549.1 | 1 - 2 | 542 | 66 | |
| GenBank mRNA | EF166088.1 | - | 1 - 2 | 542 | 66 | |
More Information
Set Membership: |
|

gnrhr
Genomic Map