Assay Details
Gene Symbol: c6orf203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114216.1 | NP_001107688.1 | 3 - 4 | 585 | 68 | |
XM_012963124.2 | XP_012818578.1 | 3 - 4 | 620 | 68 | ||
XM_012963125.2 | XP_012818579.1 | 3 - 4 | 610 | 68 | ||
XM_012963126.2 | XP_012818580.1 | 3 - 4 | 601 | 68 | ||
XM_018093935.1 | XP_017949424.1 | 3 - 4 | 1489 | 68 | ||
XM_018093936.1 | XP_017949425.1 | 3 - 4 | 584 | 68 | ||
GenBank mRNA | BC121588.1 | - | 2 - 3 | 497 | 68 | |
BC157523.1 | - | 3 - 4 | 585 | 68 | ||
CU025092.1 | - | 3 - 4 | 534 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
